Biphenyl diphenyl ether eutectic

Lieferant: Thermo Fisher Scientific
Achtung

417521000 417520010
ACRO417521000EA 30.6 EUR
ACRO417521000 ACRO417520010
Biphenyl diphenyl ether eutectic
Biphenyl diphenyl ether eutectic

Appearance: Clear colorless to yellow Liquid

Formel: C₂₄H₂₀O
Schmelzpunkt: 13 °C
Dichte: 1,056 g/cm³
Flammpunkt: 123 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00148859
CAS-Nummer: 8004-13-5
UN: 3082
ADR: 9,III

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5885 to 1.5905 (20°C, 589 nm)
GC mix of phenyl ether-biphenyl: approx 70%-30%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR