Biphenyl diphenyl ether eutectic
Lieferant: Thermo Fisher Scientific
|
Achtung
|
417521000
417520010
ACRO417521000EA
30.6
EUR
ACRO417521000
ACRO417520010
Biphenyl diphenyl ether eutectic
Biphenyl diphenyl ether eutectic
Appearance: Clear colorless to yellow Liquid
Formel:
C₂₄H₂₀O Schmelzpunkt: 13 °C Dichte: 1,056 g/cm³ Flammpunkt: 123 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00148859 CAS-Nummer: 8004-13-5 UN: 3082 ADR: 9,III |
Spezifikation Testergebnisse
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5885 to 1.5905 (20°C, 589 nm) |
GC | mix of phenyl ether-biphenyl: approx 70%-30% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...