Biphenyl-3-ol 90%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3-Hydroxybiphenyl, 3-Hydroxydiphenyl, 1-Hydroxy-3-phenylbenzol, 3-Phenylphenol

417660250 417660050
ACRO417660250EA 223 EUR
ACRO417660250 ACRO417660050
Biphenyl-3-ol 90%
Biphenyl-3-ol
Formel: C₁₂H₁₀O
Molecular Weight: 170.21 g/mol
Siedepunkt: 300 °C (760 mmHg)
Schmelzpunkt: 72…80 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002294
CAS-Nummer: 580-51-8

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow to pink to light brown Crystalline powder and/or chunks
Infrared spectrum Conforms
Melting point 72 to 80 °C
GC ≥88 %
Isomer balance mainly para-isomer


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR