Kaliumdisulfit 97%, reinst

Lieferant: Thermo Fisher Scientific
Gefahr

418270250 418275000 418270025
ACRO418270250EA 24.1 EUR
ACRO418270250 ACRO418275000 ACRO418270025
Kaliumdisulfit 97%, reinst
Kaliumdisulfit
Formel: K₂O₅S₂
Dichte: 1,2 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00167605
CAS-Nummer: 16731-55-8
EINECS: 240-795-3
Merck Index: 13,07729

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Crystalline powder
Titration Iodimetric ≥96.0 %
Arsenic (As) ≤1 ppm
Iron (Fe) ≤5 ppm
Selenium (Se) ≤1 ppm
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR