(R)-(+)-Pulegon 92% pract.
Lieferant: Thermo Fisher Scientific
418501000
418505000
ACRO418501000EA
74.8
EUR
ACRO418501000
ACRO418505000
(R)-(+)-Pulegon 92% pract.
(R)-(+)-Pulegon
Formel:
C₁₀H₁₆O Siedepunkt: 97 °C (12 mmHg) Dichte: 0,931 g/cm³ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00063000 CAS-Nummer: 89-82-7 EINECS: 201-943-2 |
Spezifikation Testergebnisse
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4850 to 1.4880 (20°C, 589 nm) |
GC | ≥90 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...