Rhodamin 6G 99%, rein

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 9-[2-(Ethoxycarbonyl)phenyl]-3,6-bis(ethylamino)-2,7-dimethylxanthyliumchlorid

419010050 419010250 419011000
ACRO419010050EA 28.6 EUR
ACRO419010050 ACRO419010250 ACRO419011000
Rhodamin 6G 99%, rein
Rhodamin 6G
Formel: C₂₈H₃₁ClN₂O₃
Siedepunkt: ∼200 °C (1013 hPa)
Schmelzpunkt: 229,4 °C
MDL: MFCD00012665
CAS-Nummer: 989-38-8
EINECS: 213-584-9
UN: 2811

Bestellen

Spezifikation Testergebnisse

Appearance Red-brown to brown Crystalline powder
Infrared spectrum Conforms
UV spectrophotometry ≥98.5 %
Enantiomeric excess ≥97.5 %
Specific optical rotation -33° to -39° (20°C, 589 nm) (c=1, CHCl3)
Titration with NaOH ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR