Rhodamin 6G Perchlorat 99%, rein Laserqualität
Lieferant: Thermo Scientific
|
Achtung
|
419030025
ACRO419030025EA
360
EUR
ACRO419030025
Rhodamin 6G Perchlorat 99%, rein Laserqualität
Rhodamin 6G Perchlorat
Formel:
C₂₈H₃₁ClN₂O₇ Molecular Weight: 543,02 g/mol Schmelzpunkt: 263…265 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013117 CAS-Nummer: 13161-28-9 EINECS: 236-099-4 |
Spezifikation Testergebnisse
Appearance | Pink to red Crystalline powder or crystals |
Infrared spectrum | Conforms |
UV spectrum | Complies |
TLC | ≥98.5 % |
UV Lambda max | 528 nm (in Methanol) |
Fluorescence max | 547 nm (in Methanol) |
Optimum lasing | 563 nm (in Methanol) |
Suitability | (for use as a laser dye) Passes test |
Molar absorption coefficient | 104000 l/(mole.cm) (Lambda max) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...