Rhodamine 101 inner salt 94%, rein

Lieferant: Thermo Scientific
Achtung

Synonyme: 9-(2-Carboxyphenyl)-2,3,6,7,12,13,16,17-octahydro-1H,5H,11H,15H-Xantheno[2,3,4-ij:5,6,7-i'j′]diquinolizin-18-ium inner salt

419060010
ACRO419060010EA 128 EUR
ACRO419060010
Rhodamine 101 inner salt 94%, rein
Rhodamine 101 inner salt

Appearance: Dark purple Crystalline powder

Formel: C₃₂H₃₀N₂O₃
Molecular Weight: 490,603 g/mol
Schmelzpunkt: 200…210 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00051383
CAS-Nummer: 116450-56-7

Bestellen

Spezifikation Testergebnisse

Appearance Dark purple Crystalline powder
Infrared spectrum Conforms
HPLC ≥93 %
UV Lambda max 567 nm (in Methanol)
Molar absorption coefficient ≥83300 l/(mol.cm) (at Lambda max)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR