Rhodamine 123 99+%, rein

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3,6-Diamino-9-(2-(methoxycarbonyl)phenyl xanthylium chloride, 3,6-Diamino-9-(2-(methoxycarbonyl)phenyl)xanthylium chloride, 2-(6-Amino-3-imino-3H-xanthen-9-yl)benzoic acid methyl ester

419091000 419095000
ACRO419091000EA 237 EUR
ACRO419091000 ACRO419095000
Rhodamine 123 99+%, rein
Rhodamine 123

Appearance: Red-brown to brown Powder

Formel: C₂₁H₁₇ClN₂O₃
Molecular Weight: 380.82 g/mol
Schmelzpunkt: 235 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012664
CAS-Nummer: 62669-70-9

Bestellen

Spezifikation Testergebnisse

Appearance Red-brown to brown Powder
Infrared spectrum Conforms
UV spectrophotometry ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR