Sodium biphenyl 20% (w/w) in Diethylenglycoldiethylether

Lieferant: Thermo Fisher Scientific
Gefahr

419423000
ACRO419423000EA 278 EUR
ACRO419423000
Sodium biphenyl 20% (w/w) in Diethylenglycoldiethylether
Sodium biphenyl

Offered as 20 x 15mL


Appearance: Dark green to blue-black Solution

Formel: C₁₂H₁₀Na
Molecular Weight: 177.202 g/mol
Siedepunkt: 188 °C
Dichte: 1.003 g/cm³
Flammpunkt: 46 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00013182
CAS-Nummer: 5137-46-2
UN: 2920
ADR: 8 II

Bestellen

Spezifikation Testergebnisse

Appearance Dark green to blue-black Solution (May partially crystallize)
Titration with HClO4 15 to 25 % w/w
Refractive index 1.5480 to 1.5500 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR