Sodium polyanetholesulphonate

Lieferant: Thermo Fisher Scientific
Achtung

419630250 419631000 419635000
ACRO419630250EA 147 EUR
ACRO419630250 ACRO419631000 ACRO419635000
Sodium polyanetholesulphonate
Sodium polyanetholesulphonate

CAS No.: 55963-78-5

Formel: (C₁₀H₁₁O₄SNa)n
Lagertemperatur: Raumtemperatur
MDL: MFCD00148427
CAS-Nummer: 55963-78-5

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow to beige to tan Powder or crystals
Infrared spectrum Conforms
pH 5.0 to 7.0 (1 % in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR