L(+)-Weinsäure ACS

Lieferant: Thermo Fisher Scientific
Gefahr

420005000 420000025
ACRO420005000EA 66.2 EUR
ACRO420005000 ACRO420000025
L(+)-Weinsäure ACS
Formel: C₄H₆O₆
Siedepunkt: 399 °C (1013 hPa)
Dichte: 1,759 g/cm³ (20 °C)
Flammpunkt: 210 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00064207
CAS-Nummer: 87-69-4
EINECS: 201-766-0
REACH: 01-2119537204-47
Merck Index: 13,09158
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4e859e52081482a351d52322717614aad450742&vid=879c292903ddee7a59d5d6b2d969c4c8fc6eac75&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance White Crystals
Infrared spectrum Conforms
Residue after ignition ≤0.02 %
Titration with NaOH ≥99.0 %
Chloride (Cl) ≤10 ppm
Oxalate (C2O4) (limit about 0.1%) passes test
Phosphate (PO4) ≤10 ppm
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤5 ppm
Sulfur compounds (as SO4) ≤0.002 %
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR