Tetrabutylammoniumdihydrogenphosphat 99% für die HPLC

Lieferant: Thermo Fisher Scientific

Synonyme: Tetra-n-butylammonium phosphate monobasic, Tetrabutylammonium phosphate monobasic, Tetra-n-butylammonium dihydrogen phosphate, IPC-TBA-P, Tetra-n-butylammonium phosphate

420160050 420160250 420161000
ACRO420160050EA 234 EUR
ACRO420160050 ACRO420160250 ACRO420161000
Tetrabutylammoniumdihydrogenphosphat 99% für die HPLC
Tetrabutylammoniumdihydrogenphosphat
Formel: C₁₆H₃₈NO₄P
Schmelzpunkt: 148…151 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00064526
CAS-Nummer: 5574-97-0

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
UV at 214 nm A: ≤0.05
Titration with NaOH ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR