Tetraethylenglycolmonomethylether 98%

Lieferant: Thermo Fisher Scientific

Synonyme: mPEG4-OH

420350050 420350250
ACRO420350050EA 80.5 EUR
ACRO420350050 ACRO420350250
Tetraethylenglycolmonomethylether 98%
Tetraethylenglycolmonomethylether
Formel: C₉H₂₀O₅
Molecular Weight: 208.25 g/mol
Siedepunkt: 158…160 °C (5 mmHg)
Flammpunkt: >110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00041756
CAS-Nummer: 23783-42-8
EINECS: 245-883-5

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4440 to 1.4460 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR