N,N,N',N'-Tetrakis(2-hydroxyethyl)ethylendiamin 99%
Lieferant: Thermo Fisher Scientific
420485000
420480010
420481000
ACRO420485000EA
131
EUR
ACRO420485000
ACRO420480010
ACRO420481000
N,N,N',N'-Tetrakis(2-hydroxyethyl)ethylendiamin 99%
Formel:
C₁₀H₂₄N₂O₄ Dichte: 1,13 g/cm³ Flammpunkt: 110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002854 CAS-Nummer: 140-07-8 EINECS: 205-396-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...