Thymolblau Natriumsalz ACS

Lieferant: Thermo Fisher Scientific

420940100 420940250 420941000
ACRO420940100EA 48.2 EUR
ACRO420940100 ACRO420940250 ACRO420941000
Thymolblau Natriumsalz ACS
Thymolblau Natriumsalz
Formel: C₂₇H₂₉NaO₅S
Dichte: 1,19 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00151093
CAS-Nummer: 62625-21-2
EINECS: 263-650-6

Bestellen

Spezifikation Testergebnisse

Appearance Dark orange to green-black to brown-black Powder
Infrared spectrum Conforms
Visual transition interval From pH 8.0 (yellow) to pH 9.2 (blue)
Clarity of solution Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR