Thymolphthalein ACS

Lieferant: Thermo Fisher Scientific

Synonyme: 2-(1-Hydroxy-1,1-di(4-hydroxy-2-methyl-5-(methylethyl)phenyl)methyl)benzoesäurelacton, 3,3-Bis(4-hydroxy-5-isopropyl-o-tolyl)phthalid

420950100 420950250 420951000 420955000
ACRO420950100EA 50.3 EUR
ACRO420950100 ACRO420950250 ACRO420951000 ACRO420955000
Thymolphthalein ACS
Thymolphthalein
Formel: C₂₈H₃₀O₄
Schmelzpunkt: 251…253 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005909
CAS-Nummer: 125-20-2
EINECS: 204-729-7
Merck Index: 12,09542

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Visual transition interval from pH 8.8 (colorless) to pH 10.5 (blue)
Clarity of solution passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR