D-α-Tocopherol 97+%

Lieferant: Thermo Scientific

Synonyme: 3,4-Dihydro-2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-2H-benzopyran-6-ol, Vitamin E Alkohol, (+)-α-Tocopherol, D(+)-α-Tocopherol, Vitamin E

421031000
ACRO421031000EA 96.6 EUR
ACRO421031000
D-α-Tocopherol 97+%
D-α-Tocopherol
Formel: C₂₉H₅₀O₂
Molecular Weight: 430,71 g/mol
Siedepunkt: 200…220 °C (,1 mmHg)
Schmelzpunkt: 2,5…3.5 °C
Flammpunkt: >110 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00072045
CAS-Nummer: 59-02-9
EINECS: 233-466-0

Bestellen

Spezifikation Testergebnisse

Appearance Clear yellow to red-brown Viscous liquid
Infrared spectrum Conforms
Refractive index 1.5050 to 1.5070 (20°C, 589 nm)
GC ≥97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR