D-γ-Tocopherol 95%

Lieferant: Thermo Scientific

Synonyme: (R)-2,7,8-Trimethyl-2-((4R,8R)-4,8,12-trimethyltridecyl)chroman-6-ol, γ-Tocopherol

421041000
ACRO421041000EA 319 EUR
ACRO421041000
D-γ-Tocopherol 95%
D-γ-Tocopherol

CAS No.: 54-28-4


Appearance: Clear light brown Viscous liquid

Formel: C₂₈H₄₈O₂
Molecular Weight: 416,69 g/mol
Siedepunkt: 200 °C (,1 mmHg)
Flammpunkt: >93 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00066529
CAS-Nummer: 54-28-4
EINECS: 200-201-5
Merck Index: 15,9655

Bestellen

Spezifikation Testergebnisse

Appearance Clear light brown Viscous liquid
Infrared spectrum Conforms
HPLC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR