D-alpha-Tocopherylacetat 97%

Lieferant: Thermo Scientific

Synonyme: Vitamin E Acetate (D-form), (+)-ɑ-Tocopherol Acetate

421060010
ACRO421060010EA 38.9 EUR
ACRO421060010
D-alpha-Tocopherylacetat 97%
D-alpha-Tocopherol acetate

Appearance: Clear yellow Viscous liquid

Formel: C₃₁H₅₂O₃
Molecular Weight: 472,75 g/mol
Flammpunkt: >110 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00072052
CAS-Nummer: 58-95-7

Bestellen

Spezifikation Testergebnisse

Appearance Clear yellow Viscous liquid
Infrared spectrum Conforms
Refractive index 1.4950 to 1.4990 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR