3,3'-Dimethylbenzidin Dihydrochorid 99%

Lieferant: Thermo Fisher Scientific
Gefahr

421131000 421135000 421130025 421130250
ACRO421131000EA 145 EUR
ACRO421131000 ACRO421135000 ACRO421130025 ACRO421130250
3,3'-Dimethylbenzidin Dihydrochorid 99%
3,3'-Dimethylbenzidin Dihydrochorid
Formel: C₁₄H₁₈Cl₂N₂
Schmelzpunkt: 130 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012960
CAS-Nummer: 612-82-8
EINECS: 210-322-5
UN: 3077
ADR: 9,III

Bestellen

Spezifikation Testergebnisse

Appearance Light beige to light grey Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥98.5 % (on dry substance)
Water ≤5 % (K.F.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR