Triethanolamin (Trolamin, TEA) 99+%
Lieferant: Thermo Fisher Scientific
Synonyme:
2,2',2''-Nitrilotriethanol, Tris-(2-hydroxyethyl)amin
421630010
421630025
421631000
421630250
ACRO421630010EA
89.4
EUR
ACRO421630010
ACRO421630025
ACRO421631000
ACRO421630250
Triethanolamin (Trolamin, TEA) 99+%
Triethanolamin (Trolamin, TEA)
Formel:
C₆H₁₅NO₃ Molecular Weight: 149.19 g/mol Siedepunkt: 360 °C (760 mmHg) Schmelzpunkt: 21 °C Dichte: 1.1245 g/cm³ Flammpunkt: 190 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002855 CAS-Nummer: 102-71-6 EINECS: 203-049-8 Merck Index: 13,09739 |
|
Spezifikation Testergebnisse
Appearance | Clear colorless to light yellow Viscous liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4830 to 1.4860 (20°C, 589 nm) |
Titration with HClO4 | ≥99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...