1,1,2-Trimethyl-1H-benz[e]indol 96%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
421900250
ACRO421900250EA
58.9
EUR
ACRO421900250
1,1,2-Trimethyl-1H-benz[e]indol 96%
1,1,2-Trimethyl-1H-benz[e]indol
Formel:
C₁₅H₁₅N Schmelzpunkt: 111…117 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00082627 CAS-Nummer: 41532-84-7 EINECS: 255-429-8 UN: 2811 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | Yellow to brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 111 to 117 °C |
HPLC | ≥95 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...