1',3',3'-Trimethyl-6-hydroxyspiro(2H-1-benzopyran-2,2'-indoline) 99%

Lieferant: Thermo Fisher Scientific
Achtung

421920050
ACRO421920050EA 169 EUR
ACRO421920050
1',3',3'-Trimethyl-6-hydroxyspiro(2H-1-benzopyran-2,2'-indoline) 99%
1',3',3'-Trimethyl-6-hydroxyspiro(2H-1-benzopyran-2,2'-indoline)

Appearance: Light brown to purple Powder

Formel: C₁₉H₁₉NO₂
Molecular Weight: 293.366 g/mol
Schmelzpunkt: 149…150 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00051350
CAS-Nummer: 23001-29-8

Bestellen

Spezifikation Testergebnisse

Appearance Light brown to purple Powder
Infrared spectrum Conforms
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR