Ferroin (1,10-Phenanthrolin-Eisen(II)sulfat-Komplex) 0.025 M in wässriger Lösung
Lieferant: Thermo Fisher Scientific
|
|
422251000
422255000
ACRO422251000EA
54.8
EUR
ACRO422251000
ACRO422255000
Ferroin (1,10-Phenanthrolin-Eisen(II)sulfat-Komplex) 0.025 M in wässriger Lösung
Ferroin (1,10-Phenanthrolin-Eisen(II)sulfat-Komplex)
Formel:
C₃₆H₂₄FeN₆SO₄ Dichte: 1,006 g/cm³ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00036428 CAS-Nummer: 14634-91-4 EINECS: 238-676-6 ADR: 9,III |
Spezifikation Testergebnisse
Appearance | Dark red Liquid |
Concentration | 0.024 to 0.026 M (w/v in water) |
Suitability | (for use as ind. for Fe) Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...