Tyramin Hydrochlorid 99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-(4-Hydroxyphenyl)ethylammoniumchlorid, 4-Hydroxyphenethylammoniumchlorid, 4-(2-Aminoethyl)phenol Hydrochlorid

422380050 422380250 422381000
ACRO422380050EA 29.9 EUR
ACRO422380050 ACRO422380250 ACRO422381000
Tyramin Hydrochlorid 99%
Tyramin Hydrochlorid
Formel: C₈H₁₂ClNO
Molecular Weight: 173.64
Siedepunkt: 269 °C (760 mmHg)
Schmelzpunkt: 271…274 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012901
CAS-Nummer: 60-19-5
EINECS: 200-462-5

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Titration Argentometric 98.5 to 101.5 % (on dry substance)
Solubility (10 % in water) Clear colorless to light yellow


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR