(R)-(-)-2-Amino-3-methyl-1-butanol 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
D(-)-2-Amino-3-methyl-1-butanol, D(-)-Valinol
422470050
ACRO422470050EA
89.3
EUR
ACRO422470050
(R)-(-)-2-Amino-3-methyl-1-butanol 98%
(R)-(-)-2-Amino-3-methyl-1-butanol
Formel:
C₅H₁₃NO Molecular Weight: 103.16 g/mol Schmelzpunkt: 32…36 °C Dichte: 0.931 g/cm³ Flammpunkt: 78 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00064297 CAS-Nummer: 4276-09-9 |
Spezifikation Testergebnisse
Appearance | White to light yellow Crystals or low melting crystalline mass |
Infrared spectrum | Conforms |
Melting point | 32 to 36 °C |
GC | ≥97.5 % |
Specific optical rotation | -15° to -18° (20 °C, 589 nm) (c=10, EtOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...