Zink(II)phthalocyanin (Sulfat-Ionophor II) 96%

Lieferant: Thermo Fisher Scientific

Synonyme: Zinc phthalocyanine, 29H,31H-Phthalocyanine, zinc complex

422750050 422750250
ACRO422750050EA 53.9 EUR
ACRO422750050 ACRO422750250
Zink(II)phthalocyanin (Sulfat-Ionophor II) 96%
Zink(II)phthalocyanin (Sulfat-Ionophor II)
Formel: C₃₂H₁₆N₈Zn
Lagertemperatur: Raumtemperatur
MDL: MFCD00041955
CAS-Nummer: 14320-04-8
EINECS: 238-262-5

Bestellen

Spezifikation Testergebnisse

Appearance Blue to purple Powder
Infrared spectrum Conforms
Loss on drying ≤5 % (105°C)
UV spectrophotometry ≥95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR