Tris(diethylamino)phosphin 95%

Lieferant: Thermo Fisher Scientific
Achtung

423080250 423080050 423081000
ACRO423080250EA 180 EUR
ACRO423080250 ACRO423080050 ACRO423081000
Tris(diethylamino)phosphin 95%
Tris(diethylamino)phosphin
Formel: C₁₂H₃₀N₃P
Siedepunkt: 111 °C (6 mmHg)
Dichte: 0,89 g/cm³
Flammpunkt: 58 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00009041
CAS-Nummer: 2283-11-6
EINECS: 218-920-8
UN: 1993
ADR: 3,III

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to yellow to light orange Liquid
Infrared spectrum Conforms
Refractive index 1.4720 to 1.4750 (20°C, 589 nm)
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR