Ammoniumfluorid 98+% ACS
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Fluoroammonium
423290050
423295000
423290025
ACRO423290050EA
21.6
EUR
ACRO423290050
ACRO423295000
ACRO423290025
Ammoniumfluorid 98+% ACS
Ammonium fluoride
Formel:
NH₄F Schmelzpunkt: 160 °C Dichte: 1,0088 to 1.0096 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011423 CAS-Nummer: 12125-01-8 EINECS: 235-185-9 UN: 2505 ADR: 6.1,III Merck Index: 13,00524 |
Spezifikation Testergebnisse
Appearance | White Crystalline powder |
Residue after ignition | ≤0.01 % |
Titration excess NaOH | ≥98.0 % |
Chloride (Cl) | ≤0.001 % |
Sulfate (SO4) | ≤0.005 % |
Iron (Fe) | ≤5 ppm |
Heavy metals (as Pb) | ≤5 ppm |
Insoluble matter | ≤0.005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...