Ammoniumheptamolybdat Tetrahydrat ACS

Lieferant: Thermo Fisher Scientific
Achtung

423310050 423315000 423311000 423310025
ACRO423310050EA 24.4 EUR
ACRO423310050 ACRO423315000 ACRO423311000 ACRO423310025
Ammoniumheptamolybdat Tetrahydrat ACS
Ammoniumheptamolybdat Tetrahydrat
Formel: (NH₄)₆Mo₇O₂₄·4H₂O
Schmelzpunkt: 190 °C
Dichte: 2,49 g/cm³
Lagertemperatur: Raumtemperatur
MDL: MFCD00167059
CAS-Nummer: 12054-85-2
EINECS: 234-722-4
Merck Index: 13,00536

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder or crystals (May have a yellow to light green hue)
Titration Complexometric 81.0 to 83.0 % (as MoO3)
Chloride (Cl) ≤0.002 %
Nitrate (NO3) (Limit about 0.003%) Passes test
Phosphate (PO4) ≤5 ppm
Sulfate (SO4) ≤0.02 %
AsO4, PO4, SiO4 (as SiO2) ≤0.001 %
Magnesium (Mg) ≤0.005 %
Potassium (K) ≤0.01 %
Sodium (Na) ≤0.01 %
Heavy metals (ICP-OES) ≤0.001 %
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR