Acridine orange (chlorid salt), hydrate ∼80% Dye, pure

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: AO, C.I. 46005, 3,6-Bis(dimethylamino)acridine hydrochloride, Basic Orange 14, N,N,N’,N’-Tetramethyl-3,6-acridinediamine hydrochloride, 3,6-Acridinediamine, Acridine orange hydrochloride

423340010
ACRO423340010EA 103 EUR
ACRO423340010
Acridine orange (chlorid salt), hydrate ∼80% Dye, pure
Acridinorange (Chloridsalz)
Formel: C₁₇H₂₀ClN₃·H₂O
Molecular Weight: 301.81 g/mol (anhy)
Schmelzpunkt: 284…287 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00150070
CAS-Nummer: 1704465-79-1
EINECS: 200-614-0

Bestellen

Spezifikation Testergebnisse

Appearance Orange to brown Powder
Infrared spectrum Conforms
Loss on drying ≤5 % (105°C)
UV spectrophotometry ≥75 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR