Ammoniumsulfat 99+% ACS

Lieferant: Thermo Fisher Scientific

Synonyme: Diammoniumsulfat, di-Ammoniumsulfat

423400010 423400250 423405000 423400050 423400251
ACRO423400010EA 51.1 EUR
ACRO423400010 ACRO423400250 ACRO423405000 ACRO423400050 ACRO423400251
Ammoniumsulfat 99+% ACS
Ammoniumsulfat
Formel: (NH₄)₂SO₄
Schmelzpunkt: 280 °C
Dichte: 1,78 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003391
CAS-Nummer: 7783-20-2
EINECS: 231-984-1
REACH: 01-2119455044-46
Merck Index: 13,00559

Bestellen

Spezifikation Testergebnisse

Appearance Colorless to white Crystalline powder or crystals
Residue after ignition ≤0.005 %
pH 5.0 to 6.0 (5 % at 25.0°C)
Titration excess NaOH ≥99.0 %
Chloride (Cl) ≤5 ppm
Nitrate (NO3) ≤0.001 %
Phosphate (PO4) ≤5 ppm
Iron (Fe) ≤5 ppm
Heavy metals (ICP-OES) ≤5 ppm
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR