Ammoniumsulfat 99+% ACS
Lieferant: Thermo Fisher Scientific
Synonyme:
Diammoniumsulfat, di-Ammoniumsulfat
423400010
423400250
423405000
423400050
423400251
ACRO423400010EA
51.1
EUR
ACRO423400010
ACRO423400250
ACRO423405000
ACRO423400050
ACRO423400251
Ammoniumsulfat 99+% ACS
Ammoniumsulfat
Formel:
(NH₄)₂SO₄ Schmelzpunkt: 280 °C Dichte: 1,78 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003391 CAS-Nummer: 7783-20-2 EINECS: 231-984-1 REACH: 01-2119455044-46 Merck Index: 13,00559 |
Spezifikation Testergebnisse
Appearance | Colorless to white Crystalline powder or crystals |
Residue after ignition | ≤0.005 % |
pH | 5.0 to 6.0 (5 % at 25.0°C) |
Titration excess NaOH | ≥99.0 % |
Chloride (Cl) | ≤5 ppm |
Nitrate (NO3) | ≤0.001 % |
Phosphate (PO4) | ≤5 ppm |
Iron (Fe) | ≤5 ppm |
Heavy metals (ICP-OES) | ≤5 ppm |
Insoluble matter | ≤0.005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...