Ammoniumthiocyanat 98% ACS

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Ammoniumsulfocyanid, Ammoniumrhodanid

423410030 423415000
ACRO423410030EA 216 EUR
ACRO423410030 ACRO423415000
Ammoniumthiocyanat 98% ACS
Ammoniumthiocyanat
Formel: NH₄SCN
Schmelzpunkt: 149 °C
Dichte: 1,3 g/cm³
Lagertemperatur: Raumtemperatur
MDL: MFCD00011428
CAS-Nummer: 1762-95-4
EINECS: 217-175-6
Merck Index: 13,00564

Bestellen

Spezifikation Testergebnisse

Appearance Colorless to white Crystals
Titration Argentimetric ≥97.5 %
Residue after ignition ≤0.025 %
pH 4.5 to 6.0 (5 % at 25.0°C)
Chloride (Cl) ≤0.005 %
Sulfate (SO4) ≤0.005 %
Iron (Fe) ≤3 ppm
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 %
Iodine consuming subst. ≤0.004 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR