Cobalt(II)chlorid Hexahydrat 98-102% ACS
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
423570050
423571000
423575000
ACRO423570050EA
49.9
EUR
ACRO423570050
ACRO423571000
ACRO423575000
Cobalt(II)chlorid Hexahydrat 98-102% ACS
Formel:
CoCl₂·6H₂O Siedepunkt: 110 °C (1013 hPa) Schmelzpunkt: 86 °C Dichte: 1,92 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149652 CAS-Nummer: 7791-13-1 EINECS: 231-589-4 UN: 3077 ADR: 9,III Merck Index: 13,02462 |
Spezifikation Testergebnisse
Appearance | Violet Crystalline powder |
Titration Complexometric | 98.0 to 102.0 % |
Nitrate (NO3) | ≤100 ppm |
Sulfate (SO4) | ≤100 ppm |
Calcium (Ca) | ≤50 ppm |
Copper (Cu) | ≤20 ppm |
Iron (Fe) | ≤50 ppm |
Magnesium (Mg) | ≤50 ppm |
Nickel (Ni) | ≤1000 ppm |
Potassium (K) | ≤100 ppm |
Sodium (Na) | ≤500 ppm |
Zinc (Zn) | ≤300 ppm |
Insoluble matter | ≤0.01 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...