Diphenylamin ACS

Lieferant: Thermo Fisher Scientific
Gefahr

423650050 423651000 423655000
ACRO423650050EA 46 EUR
ACRO423650050 ACRO423651000 ACRO423655000
Diphenylamin ACS
Diphenylamin
Formel: (C₆H₅)₂NH
Siedepunkt: 302 °C (1013 hPa)
Schmelzpunkt: 52…54 °C
Dichte: 1.165 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003014
CAS-Nummer: 122-39-4
EINECS: 204-539-4
UN: 3077
ADR: 9,III
Merck Index: 13,03349
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=23b05bdc0d251be7093ced24f3f9389bc8e8d3c5&vid=f543cfadbe22d37b18bd37917bb8b5672a6d2df4&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder or crystals or flakes
Infrared spectrum Conforms
Melting point 52.5 to 54.0 °C
Solubility (in alcohol) Passes test
Residue after ignition ≤0.03 %
Sensitivity (to nitrate) Passes test
Nitrate (NO3) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR