Bromwasserstoff ∼48% in Wasser ACS

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Bromwasserstoff, Hydrogenbromid

423785000 423780025
ACRO423785000EA 79.1 EUR
ACRO423785000 ACRO423780025
Bromwasserstoff ∼48% in Wasser ACS
Bromwasserstoff
Formel: HBr
Molecular Weight: 80.91 g/mol
Siedepunkt: -66.7
Schmelzpunkt: –11 °C
Dichte: 1.48 g/cm³
Lagertemperatur: Raumtemperatur
MDL: MFCD00011323
CAS-Nummer: 10035-10-6
UN: 1788
ADR: 8 II
Merck Index: 13,04799

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to light yellow Solution
Residue after ignition ≤0.002 %
Titration with NaOH 47.0 to 49.0 %
Chloride (Cl) ≤500 ppm
Iodide (I) ≤30 ppm
Phosphate (PO4) ≤10 ppm
SO4 and SO3 (as SO4) ≤0.003 %
Iron (Fe) ≤1 ppm
Selenium (Se) ≤0.01 ppm
Heavy metals (ICP-OES) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR