Mangan(II)sulfat Monohydrat 98+% ACS

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Mangansulfat Monohydrat

423910050 423915000 423910025
ACRO423910050EA 20.9 EUR
ACRO423910050 ACRO423915000 ACRO423910025
Mangan(II)sulfat Monohydrat 98+% ACS
Mangan(II)sulfat Monohydrat
Formel: MnSO₄·1H₂O
Molecular Weight: 169.02 g/mol
Schmelzpunkt: 700
Lagertemperatur: Raumtemperatur
MDL: MFCD00149159
CAS-Nummer: 10034-96-5
EINECS: 232-089-9
UN: 3077
ADR: 9 III
Merck Index: 13,05763

Bestellen

Spezifikation Testergebnisse

Appearance Light pink Powder
Titration Complexometric 98.0 to 101.0 %
Chloride (Cl) ≤50 ppm
Calcium (Ca) ≤50 ppm
Iron (Fe) ≤20 ppm
Magnesium (Mg) ≤50 ppm
Nickel (Ni) ≤200 ppm
Potassium (K) ≤100 ppm
Sodium (Na) ≤500 ppm
Zinc (Zn) ≤50 ppm
Heavy metals (as Pb) ≤20 ppm
Insoluble matter ≤0.01 %
KMnO4-reducing substance Passes test
Loss on ignition 10.0 % to 12.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR