N-(1-Naphthyl)ethylenediamine Dihydrochlorid 98+% ACS

Lieferant: Thermo Fisher Scientific
Achtung

423990250 423991000 423995000 423990050
ACRO423990250EA 178 EUR
ACRO423990250 ACRO423991000 ACRO423995000 ACRO423990050
N-(1-Naphthyl)ethylenediamine Dihydrochlorid 98+% ACS
N-2-Aminoethyl-1-naphthylamin Dihydrochlorid
Formel: C₁₂H₁₆Cl₂N₂
Schmelzpunkt: 200 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00012556
CAS-Nummer: 1465-25-4
EINECS: 215-981-2
Merck Index: 12,06496
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3643e6df3e9cf8dc627823e9090c97004ee25308&vid=a30fc28279c11af12c6b735b00b84fc8f08caf34&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance White to beige to pink Powder or crystals and/or chunks
Infrared spectrum Conforms
Titration Argentometric ≥98.0 % (on dry substance)
Water ≤5 % (K.F.)
Loss on drying ≤5 % (1 g, 80°C, 4 h)
Solubility Passes test
Sensitivity (to sulfanilamide) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR