Natriumhydrogencarbonat 99.7+% ACS

Lieferant: Thermo Fisher Scientific

424270010 424270250 424270050 424270251
ACRO424270010EA 44.1 EUR
ACRO424270010 ACRO424270250 ACRO424270050 ACRO424270251
Natriumhydrogencarbonat 99.7+% ACS
Natriumhydrogencarbonat
Formel: NaHCO₃
Schmelzpunkt: 270 °C
Dichte: 2,159 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003528
CAS-Nummer: 144-55-8
EINECS: 205-633-8
Merck Index: 13,08655
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6b1b80d2afb3deab0c4ca836a921fcaad637890&vid=1163b37d9cf1c10fe0ed252e02a51fe12d6b50dd&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance White Powder
Titration with HCl 99.7 to 100.3 % (On dried substance)
Chloride (Cl) ≤30 ppm
Phosphate (PO4) ≤10 ppm
Ammonium (NH4) ≤5 ppm
Calcium (Ca) ≤200 ppm
Iron (Fe) ≤10 ppm
Magnesium (Mg) ≤50 ppm
Potassium (K) ≤50 ppm
Heavy metals (as Pb) ≤5 ppm
Sulfur compounds (as SO4) ≤0.003 %
Insoluble matter ≤0.015 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR