Natriumcarbonat, wasserfrei 99.6% ACS

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: wasserfreie Soda

424285000 424280025
ACRO424285000EA 42.2 EUR
ACRO424285000 ACRO424280025
Natriumcarbonat, wasserfrei 99.6% ACS
Natriumcarbonat
Formel: Na₂CO₃
Molecular Weight: 105.99 g/mol
Siedepunkt: 1600 °C (760 mmHg)
Schmelzpunkt: 851 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003494
CAS-Nummer: 497-19-8
EINECS: 207-838-8
REACH: 01-2119485498-19
Merck Index: 14,08596
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a6ff30f1487cdef86add98b38bfa8eb3dac14f6&vid=c5b129b7acb5c897b9d970ea3130d61fb1934b54&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance White Powder or granular powder
Titration with HCl ≥99.5 % (On dried substance)
Chloride (Cl) ≤10 ppm
Phosphate (PO4) ≤10 ppm
Calcium (Ca) ≤300 ppm
Iron (Fe) ≤5 ppm
Magnesium (Mg) ≤50 ppm
Potassium (K) ≤50 ppm
Heavy metals (as Pb) ≤5 ppm
Silicon dioxide (SiO2) ≤50 ppm
Sulfur compounds (as SO4) ≤0.003 %
Insoluble matter ≤0.01 %
Loss on heating ≤1.0 % (285°C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR