Natriumdihydrogenphosphat Monohydrat 98+% ACS

Lieferant: Thermo Fisher Scientific

Synonyme: Natriumphosphat monobasisch Monohydrat

424390025 424390250 424395000 424390100
ACRO424390025EA 164 EUR
ACRO424390025 ACRO424390250 ACRO424395000 ACRO424390100
Natriumdihydrogenphosphat Monohydrat 98+% ACS
Natriumdihydrogenphosphat Monohydrat
Formel: NaH₂PO₄·H₂O
Molecular Weight: 137.99 g/mol
Schmelzpunkt: 100
Lagertemperatur: Raumtemperatur
MDL: MFCD00149208
CAS-Nummer: 10049-21-5
EINECS: 231-449-2
Merck Index: 13,08734
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4e3b73d9200cc18a9423c5b98ad4affbd4aca8a&vid=a46f80a2f4b075e731486341a7be9b01c444a160&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance Semitransparent white Crystalline powder or crystals
pH 4.1 to 4.5 (5 % solution at 25.0°C)
Titration with NaOH 98.0 to 102.0 %
Chloride (Cl) ≤5 ppm
Sulfate (SO4) ≤30 ppm
Calcium (Ca) ≤50 ppm
Iron (Fe) ≤10 ppm
Potassium (K) ≤100 ppm
Heavy metals (as Pb) ≤10 ppm
Insoluble matter ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR