Natriumwolframat Dihydrat 99+% ACS
Lieferant: Thermo Fisher Scientific
|
Achtung
|
424470050
424475000
424471000
ACRO424470050EA
25.9
EUR
ACRO424470050
ACRO424475000
ACRO424471000
Natriumwolframat Dihydrat 99+% ACS
Natriumwolframat Dihydrat
Formel:
Na₂WO₄·2H₂O Schmelzpunkt: 100 °C Dichte: 3,23 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149190 CAS-Nummer: 10213-10-2 EINECS: 236-743-4 Merck Index: 12,08847 |
Spezifikation Testergebnisse
Appearance | White to light yellow Crystalline powder or flakes |
Gravimetry | 99.0 to 101.0 % |
Chloride (Cl) | ≤50 ppm |
Sulfate (SO4) | ≤100 ppm |
Molybdenum (Mo) | ≤10 ppm |
Heavy metals & Fe (as Pb) | ≤0.001 % |
Titrable free base | ≤0.02 meq/g |
Insoluble matter | ≤0.01 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...