Thioacetamid 99+% ACS

Lieferant: Thermo Fisher Scientific
Gefahr

424530250 424531000 424535000
ACRO424530250EA 48.7 EUR
ACRO424530250 ACRO424531000 ACRO424535000
Thioacetamid 99+% ACS
Thioacetamid
Formel: CH₃CSNH₂
Schmelzpunkt: 112…114 °C
Dichte: 1,37 g/cm³
Lagertemperatur: Raumtemperatur
MDL: MFCD00008070
CAS-Nummer: 62-55-5
EINECS: 200-541-4
Merck Index: 13,09391
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe2000e62db991c85340c257ac6d64ca42c3551a&vid=18e76b2cdee9ee22a2618ec371858a498f218457&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 111 to 114 °C
Titration Argentometric ≥99.0 %
Residue after ignition ≤0.05 %
Clarity of solution (2% soln.) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR