Thioharnstoff 99+% ACS

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Aminomercaptoameisensäureamid, Thiocarbamid

424540010 424542500
ACRO424540010EA 170 EUR
ACRO424540010 ACRO424542500
Thioharnstoff 99+% ACS
Thioharnstoff
Formel: NH₂CSNH₂
Molecular Weight: 76.12 g/mol
Schmelzpunkt: 171…175 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00008067
CAS-Nummer: 62-56-6
EINECS: 200-543-5
UN: 3077
ADR: 9 III
Merck Index: 13,09443
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fff6222c54fe16ea4d36edcbf320745cb1e18d6&vid=e9ae5dc49e92d68f893244a5cd4e0245f289ed09&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 174 to 177 °C
Titration Argentometric ≥99.0 % (On dried basis)
Loss on drying ≤0.5 % (1.5 g, 105 °C, 2 h)
Solubility (16.5g in 100ml water,25 °C) Passes test
Residue after ignition ≤0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR