2-(Methylsulfonyl)anilin Hydrochlorid 95%
Lieferant: Thermo Scientific
|
Achtung
|
424840025
ACRO424840025EA
77.7
EUR
ACRO424840025
2-(Methylsulfonyl)anilin Hydrochlorid 95%
2-(Methylsulfonyl)anilin Hydrochlorid
Formel:
C₇H₁₀ClNO₂S Schmelzpunkt: 139…141 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00216484 CAS-Nummer: 205985-95-1 |
Spezifikation Testergebnisse
Appearance | White to beige Powder |
Infrared spectrum | Conforms |
HPLC | ≥94 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...