3-(Methylsulfonyl)benzoesäure 95%
Lieferant: Thermo Fisher Scientific
424860025
ACRO424860025EA
58.5
EUR
ACRO424860025
3-(Methylsulfonyl)benzoesäure 95%
3-(Methylsulphonyl)benzoic acid
Formel:
C₈H₈O₄S Molecular Weight: 200.21 g/mol Schmelzpunkt: 230…238 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00216488 CAS-Nummer: 5345-27-7 |
Spezifikation Testergebnisse
Appearance | White to light yellow Powder |
Infrared spectrum | Conforms |
Melting point | 230 to 238 °C |
HPLC | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...