3-(Methylsulfonyl)anilin Hydrochlorid 95%

Lieferant: Thermo Fisher Scientific
Achtung

424870025
ACRO424870025EA 70.9 EUR
ACRO424870025
3-(Methylsulfonyl)anilin Hydrochlorid 95%
3-(Methylsulfonyl)anilin Hydrochlorid
Formel: C₇H₁₀ClNO₂S
Schmelzpunkt: 227…230 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00216487
CAS-Nummer: 80213-28-1

Bestellen

Spezifikation Testergebnisse

Appearance Off-white to light beige Powder
Infrared spectrum Conforms
Titration Argentometric ≥94 %
Loss on drying ≤5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR