4-(Methylsulfonyl)benzonitril 95%
Lieferant: Thermo Scientific
|
Achtung
|
424890025
ACRO424890025EA
68
EUR
ACRO424890025
4-(Methylsulfonyl)benzonitril 95%
4-(Methylsulfonyl)benzonitril
Formel:
C₈H₇NO₂S Molecular Weight: 181,2100 g/mol Schmelzpunkt: 137…142 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00216489 CAS-Nummer: 22821-76-7 |
Spezifikation Testergebnisse
Appearance | White to orange to brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 137 to 142 °C |
GC | ≥94 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...